accessibility menu, dialog, popup

UserName

CAS RN 88-14-2

IUPAC Name: furan-2-carboxylic acid
Synonyms: furoic acid β-furoate β-furancarboxylate β-furancarboxylic acid acide 2-furoique kyselina 2-furoova acido 2-furoico sodium salt 2-furoic acid pyromucic acid β-furoic acid
Molecular Weight (g/mol): 112.08
Molecular Formula: C5H4O3
InChi Key: SMNDYUVBFMFKNZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CO1
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (5)
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)