missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015742
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015742 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015742 Supplier Merck Emd Millipore Supplier No. 57552100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 88-14-2
Physical Form Neat
Linear Formula C5H4O3
MDL Number MFCD00003238
Melting Point 128°C to 132°C
Boiling Point 230°C to 232°C
Synonym Furan-2-carboxylic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 112.08
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less