missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic Acid, 97-100%, Spectrum™ Chemical

Catalog No. 18608218
Change view
Click to view available options
Quantity:
100 g
25 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608218 25 kg Poly Bottle
18608217 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608218 Supplier Spectrum Chemical Mfg Cor Supplier No. F114025KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-14-2

Specifications

CAS Max % 1
Boiling Point 230 °C (446 °F)
Packaging Poly Bottle
Quantity 25 kg
Molecular Weight (g/mol) 112.08
Percent Purity 97 to 100%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 2-Furoic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.