accessibility menu, dialog, popup

UserName

H3COO(R)(R)H3C(S)(S)FO(S)H3C(R)F(S)OH(S)H3COSF

CAS RN 80474-14-2

IUPAC Name: (1R,2R,3aS,5S,9aS,9bR,10S,11aS)-5,9b-difluoro-1-{[(fluoromethyl)sulfanyl]carbonyl}-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl propanoate
Synonyms: fluticasone propionate
Molecular Weight (g/mol): 500.57
Molecular Formula: C25H31F3O5S
InChi Key: WMWTYOKRWGGJOA-ZHLGSTKJSA-N
SMILES: CCC(=O)O[C@@]1([C@H](C)C[C@H]2C3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)