missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone propionate, Thermo Scientific Chemicals

Catalog No. AC462101000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462101000 100 mg
AC462105000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462101000 Supplier Thermo Scientific Chemicals Supplier No. 462101000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80474-14-2
Molecular Formula C25H31F3O5S
Molecular Weight (g/mol) 500.57
InChI Key WMWTYOKRWGGJOA-ZHLGSTKJSA-N
PubChem CID 91820603

Specifications

Melting Point 272°C to 273°C
Quantity 100 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in dioxane
Formula Weight 500.57
Chemical Name or Material Fluticasone propionate

RUO – Research Use Only