missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone Propionate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012246
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012246 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012246 Supplier Merck Emd Millipore Supplier No. PHR1702200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Fluticasone Propionate, a neutral, highly potent trifluorinated corticosteroid, is a component of aqueous nasal sprays. It shows potent topical anti-inflammatory activity and is highly efficient in suppressing the nasal symptoms associated with rhinitis and asthma. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 80474-14-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C25H31F3O5 S
MDL Number MFCD00866007
Synonym Fluticasone propionate; (6 alpha,11 beta,16 alpha,17 alpha)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid S-(fluoromethyl) ester
Type Certified Reference Material
Quantity 200 mg
Formula Weight 500.57
Grade Certified Reference Material