missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone Propionate 98.0+%, TCI America™

Catalog No. F0525100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0525100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0525100MG Supplier TCI America Supplier No. F0525100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80474-14-2
Molecular Formula C25H31F3O5S
Molecular Weight (g/mol) 505.604
MDL Number MFCD00866007
InChI Key WMWTYOKRWGGJOA-AMTWIDTLSA-N
Synonym fluticasone propionate-d5, fluticasone propionate d5, fluticasone propionate/, fluticasone-d5 propionate
PubChem CID 91820603
IUPAC Name [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] 2,2,3,3,3-pentadeuteriopropanoate
SMILES CCC(=O)OC1(C(CC2C1(CC(C3(C2CC(C4=CC(=O)C=CC43C)F)F)O)C)C)C(=O)SCF

Specifications

Melting Point 275°C
Color White
Quantity 100 mg
Formula Weight 500.57
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fluticasone Propionate
RTECSNumber BV7980000
TSCA No