CAS RN 7791-13-1
Cobaltous Chloride Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L Synonym: Cobalt (II) Chloride IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobaltous Chloride CS, USP/EP Red Primary Solution, Ricca Chemical
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ2-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Zero Oxygen Standard, for Calibration of Dissolved Oxygen Meters, Ricca Chemical
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ2-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobalt(II) chloride hexahydrate, 99.9% (metals basis)
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobalt(II) chloride hexahydrate, ACS, 98.0-102.0%
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobalt(II) chloride hexahydrate, for analysis
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobalt(II) chloride hexahydrate, 98%
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L Synonym: Cobaltous chloride hexahydrate IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobalt(II) chloride hexahydrate, pure
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobaltous Chloride TS, 2% (w/v) in dilute Hydrochloric Acid, Ricca Chemical
Chemical Name or Material: Cobaltous Chloride Name Note: Test Solution CAS: 7791-13-1 CAS Min %: 1.9 CAS Max %: 1.98 CAS: 7732-18-5 CAS Min %: 95.0 CAS Max %: 99.0 CAS: 7647-01-0 CAS Min %: 1.12 CAS Max %: 1.17
Cobalt Chloride, 1.2% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L Synonym: Cobalt (II) Chloride,Cobalt muriate hexahydrate PubChem CID: 24643 ChEBI: CHEBI:53503 IUPAC Name: λ2-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobaltous Chloride, For Nitrogen (Ammonia), Nitrogen (Organic), LabChem™
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L Synonym: Cobalt (II) Chloride,Cobalt muriate hexahydrate PubChem CID: 24643 ChEBI: CHEBI:53503 IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cobaltous Chloride, Hexahydrate, ACS Grade, 98.0 to 102.0% , LabChem™
CAS: 7791-13-1 Molecular Formula: Cl2CoH12O6 Molecular Weight (g/mol): 237.92 MDL Number: MFCD00149652 InChI Key: GFHNAMRJFCEERV-UHFFFAOYSA-L IUPAC Name: λ²-cobalt(2+) hexahydrate dichloride SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
- 1
- 2