missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Chloride Hexahydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCC0660500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCC0660500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCC0660500 Supplier Ricca Chemical Company Supplier No. RDCC0660500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity 98 to 102%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Cobalt Chloride, Hexahydrate
ShelfLife 60 months