missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent

Catalog No. AC423575000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423575000 500 g Plastic Bottle
AC423571000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423575000 Supplier Thermo Scientific Chemicals Supplier No. 423575000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Lilac
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 98-102%
Linear Formula CoCl2·6H2O
Fieser 01,155; 06,127; 11,479
Merck Index 15, 2421
Formula Weight 237.93
Percent Purity 98.0 to 102.0%
Grade ACS Reagent
Packaging Plastic Bottle
Vapor Pressure Negligible
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement GHS P Statement May damage fertility. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Suspected of causing genetic defects. May cause cancer by inhalation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.
RTECSNumber GG0200000

RUO – Research Use Only