missing translation for 'onlineSavingsMsg'
Learn More

Zero Oxygen Standard, for Calibration of Dissolved Oxygen Meters, Ricca Chemical

Catalog No. 94204
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Clear PET-G Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
94204 120 mL Natural Poly Bottle
942016 500 mL Clear PET-G Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 94204 Supplier Ricca Chemical Company Supplier No. 94204
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7791-13-1 , 7757-83-7 , 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.0, 4.67, 0.0023
CAS Max % 97.0, 4.86, 0.0023
Physical Form Liquid
Color Colorless to Yellow
Melting Point Approximately 0°C
Quantity 120 mL
Chemical Name or Material Zero Oxygen Standard, For Calibration of Dissolved Oxygen Meters