missing translation for 'onlineSavingsMsg'
Learn More

Pt-Co/Hazen/APHA Colour Reference Standard ASTM D 1209 (ASTM colour 100), MilliporeSigma™ Supelco™

Catalog No. 111016648
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016648 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016648 Supplier Merck Emd Millipore Supplier No. 134190500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7791-13-1
Type Color Reference Standard
Quantity 500 mL
Grade Certified Reference Material