CAS RN 7778-77-0
Buffer Solution, pH 7.00, Color-Coded Yellow (Certified), Fisher Chemical™
CAS: 7778-77-0
Buffer Solution, pH 7.00 (Certified), Fisher Chemical™
Potassium Phosphate Monobasic/Sodium Hydroxide
Potassium Phosphate Monobasic (Crystalline/Certified ACS), Fisher Chemical™
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium Phosphate Monobasic (White Crystals), Fisher BioReagents
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Buffer Solution, pH 7.40 (Certified), Fisher Chemical™
Potassium Phosphate Monobasic/Sodium Hydroxide
Potassium Phosphate Monobasic (Crystalline), Fisher Chemical
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium Phosphate Monobasic (Crystalline/NF/FCC), Fisher Chemical™
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium Phosphate Monobasic (Crystalline/NF/FCC/EP/BP), Fisher Chemical
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Potassium dihydrogen phosphate, ACS, 99.0% min
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Potassium phosphate, monobasic, 99+%, ACS reagent
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M Synonym: potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate PubChem CID: 516951 ChEBI: CHEBI:63036 SMILES: [K+].OP(O)([O-])=O
Thermo Scientific Chemicals Potassium phosphate, 0.2M buffer soln., pH 7.4
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.08 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O
Thermo Scientific Chemicals Potassium phosphate, 0.2M buffer soln., pH 7.0
CAS: 7778-77-0 Molecular Formula: H2KO4P Molecular Weight (g/mol): 136.084 MDL Number: MFCD00011401 MFCD00147253 InChI Key: GNSKLFRGEWLPPA-UHFFFAOYSA-M IUPAC Name: potassium dihydrogen phosphate SMILES: [K+].OP(O)([O-])=O