missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Monobasic (Crystalline/NF/FCC/EP/BP), Fisher Chemical

Catalog No. P386500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P386500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P386500 Supplier Thermo Fisher Scientific Supplier No. P386500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Color White
pH 4.5
Physical Form Solid
Quantity 500 g
Solubility Information Pass Test (EP/BP)
Formula Weight 136.09
Percent Purity ≥98.0% (FCC)
Grade BP/EP/FCC/NF
Identification Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (NF), Pass Test (NF/FCC/EP/BP)
Loss on Drying 1.0% max. (FCC), 1.0% max. (FCC), 1.0% max. (NF), 2.0% max. (EP/BP), 1.0% max. (NF), 2.0% max. (BP), 2.0% max. (EP)
Packaging Poly Bottle
Appearance Clear and Colorless (EP/BP)
Chemical Name or Material Potassium Phosphate Monobasic
Show More Show Less