missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 6.00 (Certified), Fisher Chemical™

Catalog No. SB104500
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB104500 500 mL Poly Bottle
SB1041 1 L Poly Bottle
SB10420 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SB104500 Supplier Thermo Fisher Scientific Supplier No. FLSB104500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer Solution, pH 6.00
Melting Point 0°C
CAS 7778-77-0, 7732-18-5, 1310-73-2
Boiling Point 100°C
Color Undesignated
pH 6.00
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less