missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7.00, Color-Coded Yellow (Certified), Fisher Chemical™

Catalog No. SB107500
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB107500 500 mL Poly Bottle
SB1074 4 L Poly Bottle
SB10720 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SB107500 Supplier Thermo Fisher Scientific Supplier No. SB107 500
Edge

Specifications

Chemical Name or Material Buffer Solution, pH 7.00, Color-Coded Yellow
Absorbance Pass Test
Melting Point 0°C
CAS 7778-77-0, 1310-73-2, 7732-18-5, 1934-21-0
Boiling Point 100°C
Color Yellow
Density 1.0g/cm³
Description Potassium Phosphate Monobasic/Sodium Hydroxide. pH/temperature conversions given on label.
pH 7.00
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Specific Gravity 1
Traceability to NIST Traceable to NIST.
Vapor Pressure 760mmHg at 20°C
Solubility Information Soluble in water
Grade Certified
Show More Show Less