missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7.40 (Certified), Fisher Chemical™

Catalog No. SB1101
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB1101 1 L
SB110500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB1101 Supplier Thermo Fisher Scientific Supplier No. FLSB1101
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Phosphate Monobasic/Sodium Hydroxide

Specifications

Chemical Name or Material Buffer Solution, pH 7.40
Melting Point 0°C
CAS 7778-77-0, 7758-11-4, 7732-18-5
Boiling Point 100°C
Density 1.0g/cm³
pH 7.4
Physical Form Liquid
Quantity 1 L
Identification Pass Test
MDL Number MFCD00134130
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less