accessibility menu, dialog, popup

UserName

CAS RN 76-60-8

IUPAC Name: 3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: bromocresol green bromcresol green
Molecular Weight (g/mol): 698.01
Molecular Formula: C21H14Br4O5S
InChi Key: FRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
Molecular Weight (g/mol) 
  • (8)
  • (16)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)
  • (8)
  • (3)
Grade 
  • (10)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (4)
  • (3)
  • (3)
  • (6)
  • (1)
pH 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (4)
  • (1)
  • (1)
  • (4)
  • (4)
  • (5)
  • (9)
  • (8)
  • (16)
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)
  • (8)
  • (3)
  • (10)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
  • (6)
  • (1)
  • (6)
  • (2)
  • (1)