missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 2002464
Change view
Click to view available options
Quantity:
5 g
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2002464 5 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2002464 Supplier Avantor Supplier No. C94601

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

Formula Weight 698.02
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 5 g
Chemical Name or Material Bromocresol Green