missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green (0.04% in Water) [for pH Determination], TCI America™

Catalog No. B0579500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0579500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0579500ML Supplier TCI America Supplier No. B0579500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005874
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym bromocresol green, bromcresol green, bromo cresol green, tetrabromo-m-cresolphthalein sulfone, 3',3,5',5-tetrabromo-m-cresolsulfonephthalein, 3,3-bis 3,5-dibromo-4-hydroxy-2-methylphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, bromocresol blue, unii-8ygn0y942m, bromocresol green, acs, o-toluenesulfonic acid, alpha-hydroxy-, gamma-sultone
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

Color Green
Quantity 500 mL
Formula Weight 698.01
Physical Form Liquid
Chemical Name or Material Bromocresol Green (0.04% in Water) [for pH Determination]
RTECSNumber SJ7456000
TSCA Yes