missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, TCI America™

Catalog No. B057825G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B057825G 25 g
B05781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B057825G Supplier TCI America Supplier No. B057825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005874
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym Bromocresol Blue
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

Melting Point 219°C
pH 3.8 to 5.4
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 698.01
Grade Laboratory
Chemical Name or Material Bromocresol Green
RTECSNumber SJ7456000
TSCA Yes