Learn More
Thermo Scientific Chemicals Bromocresol Green, ACS
Description
Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.
Solubility
Soluble in water, ethanol, methyl cellosolve.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 225°C (decomposition) |
| Beilstein | 372527 |
| Merck Index | 14,1386 |
| Color | Brown to Yellow |
| Solubility Information | Soluble in water,ethanol,methyl cellosolve. |
| Quantity | 5 g |
| Physical Form | Powder |
| Formula Weight | 698.04 |
| Grade | ACS |
| Chemical Name or Material | Bromocresol Green |
Safety and Handling
| EINECSNumber | 200-972-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only