CAS RN 7558-80-7
Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™
CAS: 7558-80-7 Molecular Formula: H2NaO4P Molecular Weight (g/mol): 119.98 MDL Number: MFCD00003527,MFCD00146206 InChI Key: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate PubChem CID: 23672064 ChEBI: CHEBI:37585 SMILES: [Na+].OP(O)([O-])=O
Buffer Solution Concentrate, pH 6.00 (Certified), Fisher Chemical
Sodium Phosphate Monobasic Anhydrous (USP), Fisher Chemical
CAS: 7558-80-7 Molecular Formula: H2NaO4P Molecular Weight (g/mol): 119.98 MDL Number: MFCD00003527,MFCD00146206 InChI Key: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate PubChem CID: 23672064 ChEBI: CHEBI:37585 IUPAC Name: sodium dihydrogen phosphate SMILES: [Na+].OP(O)([O-])=O
Sodium phosphate, monobasic, 99%, for analysis, anhydrous
CAS: 7558-80-7 Molecular Formula: H2NaO4P Molecular Weight (g/mol): 119.98 MDL Number: MFCD00003527,MFCD00146206 InChI Key: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate PubChem CID: 23672064 ChEBI: CHEBI:37585 SMILES: [Na+].OP(O)([O-])=O
Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 7.5
CAS: 7558-80-7 Molecular Weight (g/mol): 119.976
Phosphate, 0.2M buffer soln., pH 6.8, Thermo Scientific Chemicals
CAS: 7558-80-7 Molecular Weight (g/mol): 119.976
Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 7.0
CAS: 7558-80-7 Molecular Weight (g/mol): 119.976
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.4
CAS: 7558-80-7 Molecular Weight (g/mol): 119.976
Sodium phosphate, monobasic, 98%, extra pure, anhydrous
CAS: 7558-80-7 Molecular Formula: H2NaO4P Molecular Weight (g/mol): 119.98 MDL Number: MFCD00003527,MFCD00146206 InChI Key: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate PubChem CID: 23672064 ChEBI: CHEBI:37585 IUPAC Name: sodium;dihydrogen phosphate SMILES: [Na+].OP(O)([O-])=O
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 6.5
CAS: 7558-80-7 Molecular Weight (g/mol): 119.976
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
CAS: 7558-80-7 Molecular Formula: H2NaO4P Molecular Weight (g/mol): 119.98 MDL Number: MFCD00003527,MFCD00146206 InChI Key: AJPJDKMHJJGVTQ-UHFFFAOYSA-M Synonym: Phosphate, Ion chromatography standard solution PubChem CID: 23672064 ChEBI: CHEBI:37585 IUPAC Name: sodium;dihydrogen phosphate SMILES: [Na+].OP(O)([O-])=O
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.5
CAS: 7558-80-7 Molecular Weight (g/mol): 119.976
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.2
CAS: 7558-80-7 Molecular Weight (g/mol): 119.976