missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.4

Catalog No. AAJ60785AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60785AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60785AK Supplier Thermo Scientific Chemicals Supplier No. J60785AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Buffer solution commonly used in biological research. A high buffering capacity reagent used in molecular biology, biochemistry, and chromatography. | CAS: 7558-80-7119.976 g/mol

Phosphate-buffer (abbreviated PB) is a buffer solution commonly used in biological research. A high buffering capacity reagent used in molecular biology, biochemistry, and chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7558-80-7
Molecular Weight (g/mol) 119.976

Specifications

Concentration 0.5M buffer soln.
Physical Form Liquid
Quantity 250 mL
Packaging Plastic Bottle
Recommended Storage Ambient Temperatures

RUO – Research Use Only