missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium Phosphate Monobasic, MilliporeSigma™ Supelco™

Catalog No. 111022632
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022632 50 g
111022631 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022632 Supplier Merck Emd Millipore Supplier No. 5207450G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For HPLC

Chemical Identifiers

CAS 7558-80-7

Specifications

pH Range 4 to 4.5 (20°C, 10% in H2O)
Purity or Quality Grade 99% to 101% (T)
Quantity 50 g

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany