missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution Concentrate, pH 6.00 (Certified), Fisher Chemical

Catalog No. SB106500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB106500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB106500 Supplier Thermo Fisher Scientific Supplier No. FLSB106500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer Solution, pH 6.00
Name Note Concentrate
Melting Point 0°C
CAS 7558-80-7, 1310-73-2, 50-00-0, 67-56-1, 7732-18-5
Boiling Point 100°C
pH 6.0
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
MDL Number MFCD00146206
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
Grade Certified
Show More Show Less