missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 7.5

Catalog No. AAJ61324K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61324K2 1 L
AAJ61324AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61324K2 Supplier Thermo Scientific Chemicals Supplier No. J61324K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7558-80-7119.976 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7558-80-7
Molecular Weight (g/mol) 119.976

Specifications

Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 1 L
Packaging Plastic Bottle
Recommended Storage Ambient Temperatures

RUO – Research Use Only