missing translation for 'onlineSavingsMsg'
Learn More

Thallium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Catalog No. PLTL22Y
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLTL22Y 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PLTL22Y Supplier SPEX CertiPrep Group LLC Supplier No. PLTL22Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Tl1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-28-0 , 7697-37-2 , 7732-18-5
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLTL2-2Y, -2T, -2X    10,000    PLTL2-3Y, -3X
Chemical Name or Material Thallium Standard
Show More Show Less
Hazard Category Causes skin irritation, serious eyeirritation. Depleted radioactive material.
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only