missing translation for 'onlineSavingsMsg'
Learn More

Thallium ICP-MS Standard, 1000 ppm Tl in 3% HNO3, Ricca Chemical

Catalog No. MSTL1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSTL1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSTL1KN100 Supplier Ricca Chemical Company Supplier No. MSTL1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-28-0 , 7697-37-2 , 7732-18-5
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Tl
CAS Min % 93.0, 4.76, 0.09
CAS Max % 97.0, 4.95, 0.1
Equivalence 1 mL = 1 mg Tl
Physical Form Liquid
Color Colorless
Quantity 100 mL
Chemical Name or Material Thallium ICP-MS Standard