missing translation for 'onlineSavingsMsg'
Learn More

Thallium ICP Standard, 10,000 ppm Tl in 5% HNO3, Ricca Chemical

Catalog No. PTL10KN500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PTL10KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PTL10KN500 Supplier Ricca Chemical Company Supplier No. PTL10KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-28-0 , 7697-37-2 , 7732-18-5
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Tl
CAS Min % 86.0, 11.57, 0.86
CAS Max % 89.0, 12.05, 0.89
Equivalence 1 mL = 10 mg Tl
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 500 mL
Chemical Name or Material Thallium ICP Standard
Show More Show Less