missing translation for 'onlineSavingsMsg'
Learn More

Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis)

Catalog No. AA008880Y
Change view
Click to view available options
Quantity:
75 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA008880Y 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA008880Y Supplier Thermo Scientific Chemicals Supplier No. 0008880Y

missing translation for 'validMainframeMsg'

CAS: 7440-28-0 | Tl | 204.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Quantity 75 mm
Odor Odorless
Weight ≈225g/150mm
Form Rod
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Thallium rod, 12.7mm (0.5 in.) dia.
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
EINECSNumber 231-138-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only