missing translation for 'onlineSavingsMsg'
Learn More

Thallium ICP Standard, 10,000 ppm Tl in 5% HNO3, Ricca Chemical

Catalog No. PTL10KN100
Change view
Click to view available options
Package: Natural LDPE; Size: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PTL10KN100 Package: Natural LDPE; Size: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PTL10KN100 Supplier Ricca Chemical Company Supplier No. PTL10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural LDPE Bottle
CAS 7440-28-0, 7697-37-2, 7732-18-5
Concentration or Composition Notes 10,000ppm Tl
CAS Min % 86.0, 11.57, 0.86
CAS Max % 89.0, 12.05, 0.89
Equivalence 1 mL = 10 mg Tl
Molecular Formula Tl
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
SMILES [Tl]
IUPAC Name thallium
Molecular Weight (g/mol) 204.38
PubChem CID 5359464
ChEBI CHEBI:30440
Chemical Name or Material Thallium ICP Standard
Show More Show Less