CAS RN 7440-02-0
Nickel AA Standard, 1000 ppm Ni in 3% HNO3, Ricca Chemical
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel wire, 0.5mm (0.02in) dia, annealed, 99.98% (metals basis)
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nitinol tension coil spring, Alloy M, Thermo Scientific™
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel on silica-alumina, catalyst
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis), Thermo Scientific™
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel wire, 0.15mm (0.006in) dia, annealed, Nickel 200, approximately 99% (metals basis)
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel Thinfoil, 0.01mm (0.0004in) thick, 99.95% (metals basis)
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel Thinfoil, 0.0125mm (0.000492in) thick, 99% (metals basis excluding Co), Co nominal 0.8%
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Puratronic™, 99.995% (metals basis)
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel gauze, 20 mesh woven from 0.18mm (0.007in) dia wire
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel, 99.9%, (trace metal basis), powder, -325 mesh, Thermo Scientific Chemicals
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel pieces, 5x5cm (2.0x2.0in), 99.99% (metals basis)
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.98% (metals basis)
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.69 MDL Number: MFCD00011137 MFCD06798735 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N Synonym: raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]