missing translation for 'onlineSavingsMsg'
Learn More

Nickel, 99.995%, (trace metal basis), powder, Thermo Scientific Chemicals

Catalog No. AC436780500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436780500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436780500 Supplier Thermo Scientific Chemicals Supplier No. 436780500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Boiling Point 2730.0°C
Melting Point 1455.0°C
Quantity 50 g
Assay Percent Range 99.995%
Merck Index 15,6583
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute hno3
Formula Weight 58.7
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Nickel
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instruct
EINECSNumber 231-111-4

RUO – Research Use Only