missing translation for 'onlineSavingsMsg'
Learn More

Nickel AA Standard, 1000 ppm Ni in 3% HNO3, Ricca Chemical

Catalog No. ANI1KN500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ANI1KN500 500 mL Natural Poly Bottle
ANI1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ANI1KN500 Supplier Ricca Chemical Company Supplier No. ANI1KN500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-02-0 , 7732-18-5
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Ni
CAS Min % 93.0, 4.76, 0.09
CAS Max % 97.0, 4.95, 0.1
Equivalence 1 mL = 1 mg Ni
Physical Form Liquid
Color Green
pH < 2
Melting Point Approximately 0°C
Type Single-element Standard Solution
Quantity 500 mL
Grade High Purity
Chemical Name or Material Nickel AA Standard
Show More Show Less