missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Trihydrate Certified, Ricca Chemical

Catalog No. RDCM070025F
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCM070025F 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCM070025F Supplier Ricca Chemical Company Supplier No. RDCM070025F1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified grade Methylene Blue Trihydrate suitable for general laboratory reagent applications.

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
PubChem CID 104827
IUPAC Name 3,7-bis(dimethylamino)-5λ4-phenothiazin-5-ylium trihydrate chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Dark Green
Quantity 25 g
Physical Form Powder
Grade Laboratory
Chemical Name or Material Methylene Blue, Trihydrate, Certified
ShelfLife 60 months