missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue Solution I, 0.05 mL (1 drop) = 1 mg/L Sulfide, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Methylene Blue Solution I |
| Name Note | 0.05mL (1 drop) = 1mg/L Sulfide |
| CAS | 7220-79-3, 7732-18-5 |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Quantity | 1 L |