missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagents

Catalog No. 01336762
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336762 100 g
01336761 10 g
01336760 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336762 Supplier Labchem Inc Supplier No. C6110500100C2

Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Methylene Blue Chloride Trihydrate (Basic Blue 9)
CAS 7220-79-3
Molecular Formula C16H18ClN3S, 3H2O
Synonym 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015
Molecular Weight (g/mol) 373.8981
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0001
Color Blue/Green
Quantity 100 g
Physical Form Solid
Show More Show Less