missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Solution I, 0.05 mL (1 drop) = 1 mg/L Sulfide, Ricca Chemical

Catalog No. 490516
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
490516 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 490516 Supplier Ricca Chemical Company Supplier No. 490516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Methylene Blue Solution I
CAS 7220-79-3, 7732-18-5
CAS Min % 98.0, 0.09
CAS Max % 100.0, 0.1
Packaging Natural Poly Bottle
Color Blue
Quantity 500 mL