missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, 0.05% (w/v), Ricca Chemical

Catalog No. 48404
Change view
Click to view available options
Quantity:
120 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
48404 120 mL Natural Poly Bottle
484032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48404 Supplier Ricca Chemical Company Supplier No. 48404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Methylene Blue
CAS 7220-79-3, 7732-18-5
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Concentration 0.05% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Blue
Quantity 120 mL
Physical Form Liquid
Percent Purity 0.05% W/V
Grade Laboratory
Show More Show Less