accessibility menu, dialog, popup

UserName

∗OSi∗MeMen

CAS RN 63148-62-9

IUPAC Name: Polydimethylsiloxane
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2H6OSi)n
InChi Key: —
SMILES: C[Si](C)(-*)O-*
Quantity 
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Grade 
  • (1)
  • (1)
  • (2)
  • (11)
Physical Form 
  • (1)
  • (3)
  • (4)
Boiling Point 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (19)
  • (13)
  • (2)
  • (3)
  • (13)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (11)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)