missing translation for 'onlineSavingsMsg'
Learn More

Antifoam A, Spectrum™ Chemical

Catalog No. 18600727
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600727 20 L Poly Bottle
18600723 100 mL Poly Bottle
18600724 500 mL Poly Bottle
18600725 4 L Poly Bottle
18600726 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18600727 Supplier Spectrum Chemical Mfg Cor Supplier No. A130220LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A1302

Spectrum™ Chemical Antifoam A is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00132673
IUPAC Name Polydimethylsiloxane
SMILES C[Si](C)(-*)O-*

Specifications

CAS Max % 80% to 100%, 10% to 20%, 5% to <10%, 3% to <5%, 1% to <3%
Quantity 20 L
Packaging Poly Bottle
Grade Ungraded
Chemical Name or Material Antifoam A