missing translation for 'onlineSavingsMsg'
Learn More

Poly(dimethylsiloxane) for GPC 311, MilliporeSigma™ Supelco™

Catalog No. 111014975
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014975 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014975 Supplier Merck Emd Millipore Supplier No. 81153100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.

Specifications

CAS 63148-62-9
Vapor Pressure 5 mmHg (20°C)
Flash Point 321°C
Physical Form Neat
Linear Formula [Si(CH3)2 O]n
MDL Number MFCD00084411
Synonym Dimethylpolysiloxane
Type Analytical Standard
Quantity 100 mg
Grade GPC