missing translation for 'onlineSavingsMsg'
Learn More

Silicone Fluid, 1000, Spectrum™ Chemical

Catalog No. 18606221
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly-Coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606221 1 kg Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606221 Supplier Spectrum Chemical Mfg Cor Supplier No. SI1141KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SI114, 63148-62-9

Spectrum™ Chemical Silicone Fluid, 1000

Chemical Identifiers

CAS 63148-62-9
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00132673
IUPAC Name Polydimethylsiloxane
SMILES C[Si](C)(-*)O-*

Specifications

CAS Max % 1
Quantity 1 kg
Packaging Poly-Coated Glass Bottle
Viscosity 950-1050 cSt
Grade Ungraded
Chemical Name or Material Silicone Fluid, 1000