missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicone Fluid, 100, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 440 lb. |
| Viscosity | 95.0 - 105.0 cs |
| Grade | Ungraded |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Silicone Fluid, 100 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.