missing translation for 'onlineSavingsMsg'
Learn More

Poly(dimethylsiloxane), Average Mw 95,000 (Typical), Average Mn 50,000 (Typical), MilliporeSigma™ Supelco™

Catalog No. 111014974
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014974 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014974 Supplier Merck Emd Millipore Supplier No. 18184625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.

Specifications

CAS 63148-62-9
Vapor Pressure 5 mmHg (20°C)
Flash Point 316°C
Physical Form Neat
Linear Formula [Si(CH3)2 O]n
MDL Number MFCD00084411
Synonym Dimethylpolysiloxane
Type Analytical Standard
Quantity 25 g
Grade Analytical Standard