accessibility menu, dialog, popup

UserName

CH3BrHOCH3OBrSOOO(E)

CAS RN 62625-30-3

IUPAC Name: 2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Synonyms: 2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
Molecular Weight (g/mol): 539.21
Molecular Formula: C21H15Br2O5S
InChi Key: HJZFRHKAXRLQSG-XMHGGMMESA-M
SMILES: CC1=CC(=CC(Br)=C1O)C(=C1\C=C(C)C(=O)C(Br)=C1)\C1=CC=CC=C1S([O-])(=O)=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Color 
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (12)
Grade 
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (3)
  • (4)
  • (1)
  • (8)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (12)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)