missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocrésol, sel sodique violet, qualité réactif, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Bromocresol Purple Sodium Salt |
| Point de fusion | 255 °C |
| CAS % maximal | 100 |
| Désignation | Test de réussite |
| Emballage | Verre ambré |
| Formule linéaire | C21H15BrNaO5S |
| Renseignements sur la décomposition | bromure d’hydrogène; Monoxyde de carbone; Le dioxyde de carbone; Composés soufrés |
| Couleur | Brun/Rose |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Quantité | 10 g |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust. Use only outdoors or in a well-ventilated area. Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. Take off contaminated clothing and wash before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Store locked up in a well-ventilated place. Keep container tightly closed. Dispose of contents/container to comply with local, state and federal regulations. |
| EINECSNumber | 263-655-3 |
| Recommended Storage | Température ambiante |