accessibility menu, dialog, popup

UserName

HOOOHOHHOHO

CAS RN 59-23-4

IUPAC Name: 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: galactose d-galactose glucose oxidase mannose D-hexose D-hexoses
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: WQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILES: OCC1OC(O)C(O)C(O)C1O
Percent Purity 
  • (1)
  • (6)
  • (4)
Physical Form 
  • (4)
  • (6)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (6)
  • (4)
  • (4)
  • (6)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)