missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D-Galactose

Catalog No. 3455100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3455100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3455100GM Supplier MilliporeSigma™ Supplier No. 3455100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aldo hexose

An aldo hexose

Specifications

CAS 59-23-4
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 180.2g/mol
Purity ≥97% by HPLC
Quantity 100 g
Form Powder